Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Human Nectin-4 Alexa Fluor 488-conjugated Antibody, R&D Systems™
GREENER_CHOICE

Catalog No. FAB2659G
Change view
Click to view available options
Quantity:
100 Tests
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
FAB2659G 100 Tests
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. FAB2659G Supplier R&D Systems Supplier No. FAB2659G
Add to Cart
Add to Cart
This item is not returnable. View return policy

Mouse Monoclonal Antibody

Nectin-4/PVRL4 Monoclonal specifically detects Nectin-4/PVRL4 in Human samples. It is validated for Flow Cytometry.

Specifications

Antigen Nectin-4/PVRL4
Applications Flow Cytometry
Classification Monoclonal
Clone 337516
Conjugate Alexa Fluor 488
Dilution Flow Cytometry 5 uL/10^6 cells
Formulation Supplied in a saline solution containing BSA and Sodium Azide. with Sodium Azide
Gene Accession No. Q96NY8
Gene Alias EDSS1, Ig superfamily receptor LNIR, LNIR, LNIRnectin 4, nectin-4, Nectin4, poliovirus receptor-related 4, poliovirus receptor-related protein 4, PRR4, PRR4DKFZp686K05193, PVRL4
Gene Symbols PVRL4
Host Species Mouse
Immunogen Mouse myeloma cell line NS0-derived recombinant human Nectin-4 Gly27-Val351 Accession # Q96NY8
Purification Method Affinity Purified
Quantity 100 Tests
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 81607
Test Specificity Detects human Nectin-4 in direct ELISAs and Western blots. In direct ELISAs and Western blots, does not cross-react with recombinant human Nectin-1, -2, -3, or recombinant mouse Nectin-4.
Target Species Human
Content And Storage Protect from light. Do not freeze. 12 months from date of receipt, 2 to 8 degreesC as supplied.
Isotype IgG2b
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.