Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

R&D Systems™ Human Nectin-4 DuoSet ELISA
SDP

Catalog No. DY2659 Shop All Novus Biologicals Products
Change view
Click to view available options
Quantity:
1 Kit (for 15 Plates)
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
DY2659 1 Kit (for 15 Plates)
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. DY2659 Supplier R&D Systems™ Supplier No. DY2659
Save 15% on Immunology Workflow Reagents
Use promo code IMMUNO15
View all promotions
Add to Cart
Add to Cart
This item is not returnable. View return policy

Human Nectin-4 DuoSet ELISA from the most referenced ELISA manufacturer. Highly validated for accurate quantitation and long-term reproducibility.

The Human Nectin-4 DuoSet ELISA is a Solid Phase Sandwich ELISA that quantifies human Nectin-4/PVRL4.

Specifications

Quantity 1 Kit (for 15 Plates)
Species Human
Conjugate Biotin
Assay Range 93.80 to 6000 pg/mL
For Use With (Application) ELISA
Gene ID (Entrez) 81607
Synonym EDSS1, Ig superfamily receptor LNIR, LNIR, LNIRnectin 4, nectin-4, Nectin4, poliovirus receptor-related 4, poliovirus receptor-related protein 4, PRR4, PRR4DKFZp686K05193, PVRL4
Content And Storage Store the unopened product at 2°C to 8°C. Do not use past expiration date.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.