Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human Nephrin Antibody, R&D Systems™

Sheep Polyclonal Antibody
Supplier: R&D Systems AF4269SP
This item is not returnable.
View return policy
Description
Nephrin Polyclonal specifically detects Nephrin in Human samples. It is validated for Western Blot, Simple Western, Immunohistochemistry.Specifications
Nephrin | |
Polyclonal | |
Unconjugated | |
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
CNF, nephrin, nephrosis 1, congenital, Finnish type (nephrin), NPHNCNF, NPHS1, Renal glomerulus-specific cell adhesion receptor | |
Sheep | |
Affinity Purified | |
RUO | |
4868 | |
Reconstitute at 0.2 mg/mL in sterile PBS. | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
Western Blot, Immunohistochemistry | |
LYOPH | |
Western Blot 1 ug/mL, Simple Western 20 ug/mL, Immunohistochemistry 1-15 ug/mL | |
O60500 | |
NPHS1 | |
Mouse myeloma cell line NS0-derived recombinant human Nephrin Gln23-Thr1029 Accession # O60500 | |
25 μg | |
Primary | |
Detects human Nephrin in direct ELISAs and Western blots. In direct ELISAs, less than 20% cross-reactivity with recombinant mouse Nephrin is observed. | |
Human | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction