Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human Nephrin Antibody, R&D Systems™

Rabbit Monoclonal Antibody
$186.30 - $514.50
Specifications
Antigen | Nephrin |
---|---|
Clone | 2795E |
Dilution | Immunohistochemistry 3-25 ug/mL |
Applications | Immunohistochemistry |
Classification | Monoclonal |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
MAB42693SP
![]() |
R&D Systems
MAB42693SP |
25 μg |
Each for $186.30
|
|
|||||
MAB42693100
![]() |
R&D Systems
MAB42693100 |
100 μg |
Each for $514.50
|
|
|||||
Description
Nephrin Monoclonal antibody specifically detects Nephrin in Human samples. It is validated for ImmunohistochemistrySpecifications
Nephrin | |
Immunohistochemistry 3-25 ug/mL | |
Monoclonal | |
Purified | |
RUO | |
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. | |
CNF, nephrin, nephrosis 1, congenital, Finnish type (nephrin), NPHNCNF, NPHS1, Renal glomerulus-specific cell adhesion receptor | |
Mouse myeloma cell line NS0-derived human Nephrin protein, Gln23-Thr1029, Accession # O60500 | |
Primary | |
Reconstitute at 0.5 mg/mL in sterile PBS. |
2795E | |
Immunohistochemistry | |
Unconjugated | |
Rabbit | |
Human | |
O60500 | |
4868 | |
IgG | |
Protein A or G purified from cell culture supernatant | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 °C as supplied. 1 month, 2 to 8 °C under sterile conditions after reconstitution. 6 months, -20 to -70 °C under sterile conditions after reconstitution. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title