Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Human Nephrin Biotinylated Antibody, R&D Systems™
GREENER_CHOICE

Catalog No. BAF4269
Change view
Click to view available options
Quantity:
50 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BAF4269 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. BAF4269 Supplier R&D Systems Supplier No. BAF4269
Add to Cart
Add to Cart
This item is not returnable. View return policy

Sheep Polyclonal Antibody

Nephrin Polyclonal specifically detects Nephrin in Human samples. It is validated for Western Blot.

Specifications

Antigen Nephrin
Applications Western Blot
Classification Polyclonal
Concentration LYOPH
Conjugate Biotin
Dilution Western Blot 0.1 ug/mL
Gene Accession No. O60500
Gene Alias CNF, nephrin, nephrosis 1, congenital, Finnish type (nephrin), NPHNCNF, NPHS1, Renal glomerulus-specific cell adhesion receptor
Gene Symbols NPHS1
Host Species Sheep
Immunogen Mouse myeloma cell line NS0-derived recombinant human Nephrin Gln23-Thr1029 Accession # O60500
Purification Method Affinity Purified
Quantity 50 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 4868
Test Specificity Detects human Nephrin in Western blots. In Western blots, approximately 20% cross-reactivity with recombinant mouse Nephrin is observed.
Reconstitution Reconstitute at 0.2 mg/mL in sterile PBS.
Target Species Human
Content And Storage Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.