Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human Neprilysin-2/MMEL1 Antibody, R&D Systems™

Goat Polyclonal Antibody
Supplier: R&D Systems AF2340SP
This item is not returnable.
View return policy
Description
Neprilysin-2/MMEL1 Polyclonal specifically detects Neprilysin-2/MMEL1 in Human samples. It is validated for Western Blot, Flow Cytometry, Immunoprecipitation, CyTOF-ready.Specifications
Neprilysin-2/MMEL1 | |
Polyclonal | |
Unconjugated | |
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
EC 3.4.24, EC 3.4.24.11, MELL1, membrane metallo-endopeptidase-like 1, Membrane metallo-endopeptidase-like 2MGC119455, MGC119454, MMEL1, MMEL2, NEP2, NEP2(m), NEPIIMGC119456, Neprilysin II, neprilysin-2, Neprilysin2, NL2, NL2NL1, SEP, soluble secreted endopeptidase, zinc metallopeptidase | |
Goat | |
Affinity Purified | |
RUO | |
79258 | |
Reconstitute at 0.2 mg/mL in sterile PBS. | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
Western Blot, Flow Cytometry, Immunoprecipitation, CyTOF | |
LYOPH | |
Western Blot 0.1 ug/mL, Flow Cytometry 2.5 ug/10^6 cells, Immunoprecipitation 25 ug/mL, CyTOF-ready | |
AAL08942 | |
MMEL1 | |
Mouse myeloma cell line NS0-derived recombinant human Neprilysin-2/MMEL1 Gly69-Trp770 Accession # AAL08942 | |
25 μg | |
Primary | |
Detects human Neprilysin-2/MMEL1 in direct ELISAs and Western blots. In direct ELISAs, approximately 5% cross-reactivity with recombinant human (rh) Neprilysin is observed and less than 1% cross-reactivity with rhECE-1 and rhECE-2 is observed. | |
Human | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction