Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human Nesfatin-1/Nucleobindin-2 Antibody, R&D Systems™

Sheep Polyclonal Antibody
Supplier: R&D Systems AF5949
This item is not returnable.
View return policy
Description
Nesfatin-1/Nucleobindin-2 Polyclonal specifically detects Nesfatin-1/Nucleobindin-2 in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry.Specifications
| Nesfatin-1/Nucleobindin-2 | |
| Polyclonal | |
| Unconjugated | |
| P80303 | |
| NUCB2 | |
| E. coli-derived recombinant human Nesfatin-1/Nucleobindin-2 Pro26-Leu106 Accession # P80303 | |
| 100 μg | |
| Primary | |
| Detects human Nesfatin-1/Nucleobindin-2 in direct ELISAs and Western blots. | |
| Human | |
| IgG |
| Western Blot, Immunohistochemistry, Immunocytochemistry | |
| LYOPH | |
| Western Blot 1 ug/mL, Immunohistochemistry 5-15 ug/mL, Immunocytochemistry 5-15 ug/mL | |
| DNA-binding protein NEFA, NEFA, NEFAGastric cancer antigen Zg4, Nesfatin1, NUCB2, nucleobindin 2, nucleobindin-2, nucleobinding 2 | |
| Sheep | |
| Affinity Purified | |
| RUO | |
| 4925 | |
| Reconstitute at 0.2 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction