Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abnova™ Human NETO2 Full-length ORF (AAH12381.1, 1 a.a. - 148 a.a.) Recombinant Protein with GST-tag at N-terminal

Catalog No. 89954008
Click to view available options
Quantity:
10 μg
25 μg

Used for AP, Array, ELISA, WB-Re

This gene encodes a predicted transmembrane protein containing two extracellular CUB domains followed by a low-density lipoprotein class A (LDLa) domain. It also has an intracellular FXNPXY-like motif, which has been shown in other proteins to be essential for the internalization of clathrin coated pits during endocytosis. Alternatively spliced transcript variants have been observed, but they have not been fully characterized. [provided by RefSeq]

Sequence: MACKTAFNKTGFQEVFDPPHYELFSLRDKEISADLADLSEELDNYQKMRRSSTASRCIHDHHCGSQASSVKQSRTNLSSMELPFRNDFAQPQPMKTFNSTFKKSSYTFKQGHECPEQALEDRVMEEIPCEIYVRGREDSAQASISIDF

Specifications

Accession Number AAH12381.1
For Use With (Application) Antibody Production, Protein Array, ELISA, Western Blot
Formulation 50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
Gene ID (Entrez) 81831
Molecular Weight (g/mol) 43.4kDa
Name NETO2 (Human) Recombinant Protein (P01)
Purification Method Glutathione Sepharose 4 Fast Flow
Quality Control Testing 12.5% SDS-PAGE Stained with Coomassie Blue.
Quantity 10 μg
Immunogen MACKTAFNKTGFQEVFDPPHYELFSLRDKEISADLADLSEELDNYQKMRRSSTASRCIHDHHCGSQASSVKQSRTNLSSMELPFRNDFAQPQPMKTFNSTFKKSSYTFKQGHECPEQALEDRVMEEIPCEIYVRGREDSAQASISIDF
Storage Requirements Store at -80°C. Aliquot to avoid repeated freezing and thawing.
Regulatory Status RUO
Gene Alias FLJ10430/FLJ14724/FLJ90456/NEOT2
Common Name NETO2
Gene Symbol NETO2
Species Wheat Germ (in vitro)
Recombinant Recombinant
Protein Tag GST
Expression System wheat germ expression system
Form Liquid
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.