Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human Netrin-4 Antibody, R&D Systems™

Mouse Monoclonal Antibody
$200.50 - $655.00
Specifications
Antigen | Netrin-4 |
---|---|
Clone | 191633 |
Concentration | LYOPH |
Dilution | Western Blot 1 ug/mL |
Applications | Western Blot |
Description
Netrin-4 Monoclonal specifically detects Netrin-4 in Human samples. It is validated for Western Blot.Specifications
Netrin-4 | |
LYOPH | |
Western Blot | |
Unconjugated | |
Mouse | |
Human | |
Q9HB63 | |
59277 | |
Mouse myeloma cell line NS0-derived recombinant human Netrin-4 Val26-Lys628 Accession # Q9HB63 | |
Primary | |
Reconstitute at 0.5 mg/mL in sterile PBS. | |
Detects human Netrin-4 in direct ELISAs and Western blots. In direct ELISAs and Western blots, no cross-reactivity with recombinant mouse (rm) Netrin-1, recombinant chicken (rch) Netrin-1, rchNetrin-2, or rmNetrin-G1a is observed. |
191633 | |
Western Blot 1 ug/mL | |
Monoclonal | |
Purified | |
RUO | |
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
beta-netrin, hepar-derived netrin-like protein, netrin 4, Netrin4, NTN4, PRO3091 | |
NTN4 | |
IgG2a | |
Protein A or G purified from hybridoma culture supernatant | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title