Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human Netrin-G1a Alexa Fluor™ 647-conjugated Antibody, R&D Systems™

Mouse Monoclonal Antibody
Supplier: R&D Systems FAB10019R100UG
This item is not returnable.
View return policy
Description
Netrin-G1a Monoclonal specifically detects Netrin-G1a in Human samples. It is validated for Flow Cytometry.Specifications
Netrin-G1a | |
Monoclonal | |
Alexa Fluor 647 | |
Supplied 0.2 mg/mL in a saline solution containing BSA and Sodium Azide. | |
Mouse | |
Protein A or G purified from hybridoma culture supernatant | |
RUO | |
22854 | |
Protect from light. Do not freeze. 12 months from date of receipt, 2 to 8 °C as supplied. | |
IgG1 |
Flow Cytometry | |
1010035 | |
Flow Cytometry 0.25-1 ug/10^6 cells | |
Q9Y212 | |
Human embryonic kidney cell, HEK293-derived human Netrin-G1a, His29-Gly503, Accession # Q9Y212 | |
100 μg | |
Primary | |
Human | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction