Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Human NEU-1/Sialidase-1 Antibody, R&D Systems™
GREENER_CHOICE

Catalog No. MAB6860
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MAB6860 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. MAB6860 Supplier R&D Systems Supplier No. MAB6860
Add to Cart
Add to Cart
This item is not returnable. View return policy

Mouse Monoclonal Antibody

NEU-1/Sialidase-1 Monoclonal specifically detects NEU-1/Sialidase-1 in Human samples. It is validated for Western Blot, Immunoprecipitation.

Specifications

Antigen NEU-1/Sialidase-1
Applications Western Blot, Immunoprecipitation
Classification Monoclonal
Clone 688215
Concentration LYOPH
Conjugate Unconjugated
Dilution Western Blot 2 ug/mL, Immunoprecipitation 25 ug/mL
Formulation Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative
Gene Accession No. Q99519
Gene Alias Acetylneuraminyl hydrolase, EC 3.2.1.18, exo-alpha-sialidase, FLJ93471, G9 sialidase, Lysosomal sialidase, N-acetyl-alpha-neuraminidase 1, NANH, NEU, NEU1, SIAL1, sialidase 1 (lysosomal sialidase), sialidase-1
Gene Symbols NEU1
Host Species Mouse
Immunogen S. frugiperda insect ovarian cell line Sf 21-derived recombinant human NEU-1/Sialidase-1 Lys46-Leu415 Accession # Q99519
Purification Method Protein A or G purified from hybridoma culture supernatant
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 4758
Test Specificity Detects human NEU-1/Sialidase-1 in direct ELISAs.
Reconstitution Sterile PBS to a final concentration of 0.5 mg/mL.
Target Species Human
Content And Storage Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Form Purified
Isotype IgG2b
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.