Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Human Neuregulin-1/NRG1 Antibody, R&D Systems™
GREENER_CHOICE

Catalog No. MAB377SP
Change view
Click to view available options
Quantity:
100 μg
25 μg
500 μg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MAB377SP 25 μg
MAB377100 100 μg
MAB377500 500 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Catalog No. MAB377SP Supplier R&D Systems Supplier No. MAB377SP
Add to Cart
Add to Cart
This item is not returnable. View return policy

Mouse Monoclonal Antibody has been used in 9 publications

Neuregulin-1/NRG1 Monoclonal specifically detects Neuregulin-1/NRG1 in Human samples. It is validated for Western Blot.

Specifications

Antigen Neuregulin-1/NRG1
Applications Western Blot
Classification Monoclonal
Clone 147724
Conjugate Unconjugated
Dilution Western Blot 1 ug/mL
Formulation Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative
Gene Accession No. AAA58639
Gene Alias ARIA, GGF2, GGFglial growth factor, Heregulin-1, HGL, HGLneu differentiation factor, HRG, HRG1, HRG1-alpha, HRG1-beta 1, HRGAneuregulin 1 type IV beta 1a, MST131, MSTP131, NDFheregulin, alpha (45kD, ERBB2 p185-activator), neuregulin 1, neuregulin 1 type IV beta 3, Neuregulin1, NRG1, pro-neuregulin-1, membrane-bound isoform, pro-NRG1, sensory and motor neuron derived factor, SMDF
Gene Symbols NRG1
Host Species Mouse
Immunogen E. coli-derived recombinant human Neuregulin-1/NRG1 Ser2-Lys246 Accession # AAA58639
Purification Method Protein A or G purified from hybridoma culture supernatant
Quantity 25 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 3084
Test Specificity Detects human Neuregulin-1/NRG1 in direct ELISAs and Western blots. In direct ELISAs, no cross-reactivity with the EGF-like domains of recombinant human (rh) NRG1-beta 1 (aa 176-246) or rhNRG1-alpha (aa 177-241). Therefore, this antibody recognizes an epitope located within aa 2-176 segment N-terminal to the EGF-like domain. This segment is present in all NRG1-alpha , beta and gamma isoforms and in NDF43 and glial growth factor (GGF). It is absent from the NRG1 gene products GGF2 and SMDF.
Reconstitution Reconstitute at 0.5 mg/mL in sterile PBS.
Target Species Human
Content And Storage Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Form Purified
Isotype IgG2b
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.