Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Human Neuroligin 4X/NLGN4X Antibody, R&D Systems™
GREENER_CHOICE

Catalog No. AF5158
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AF5158 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. AF5158 Supplier R&D Systems Supplier No. AF5158
Add to Cart
Add to Cart
This item is not returnable. View return policy

Sheep Polyclonal Antibody

Produced in sheep immunized with purified, E. coli-derived, recombinant human Neuroligin 4 extracellular domain (rhNLGN4; R&D Systems, Catalog # 5158-NL). Human NLGN4 specific IgG was purified by human NLGN4 affinity chromatography.

Neuroligin 4X/NLGN4X Polyclonal specifically detects Neuroligin 4X/NLGN4X in Human samples. It is validated for Western Blot, Immunohistochemistry.

Specifications

Antigen Neuroligin 4X/NLGN4X
Applications Western Blot, Immunohistochemistry
Classification Polyclonal
Concentration LYOPH
Conjugate Unconjugated
Dilution Western Blot 1 ug/mL, Immunohistochemistry 5-15 ug/mL
Gene Accession No. Q8N0W4
Gene Alias ASPGX2, AUTSX2, HLNX, HNL4X, HNLX, KIAA1260AUTSX2, MGC22376, neuroligin 4, X-linked, NL4, NLGN, NLGN4, NLGN4neuroligin 4, NLGN4X, X-linked
Gene Symbols NLGN4X
Host Species Sheep
Immunogen E. coli-derived recombinant human Neuroligin 4/NLGN4 Met618-Thr673 Accession # Q8N0W4
Purification Method Affinity Purified
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 57502
Test Specificity Detects human Neuroligin 4/NLGN4 in direct ELISAs and Western blots. In direct ELISAs, less than 5% cross-reactivity with recombinant human (rh) NLGN3 is observed.
Reconstitution Reconstitute at 0.2 mg/mL in sterile PBS.
Target Species Human
Content And Storage Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.