Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Human Neuromedin BR/NMBR Alexa Fluor 700-conjugated Antibody, R&D Systems™
GREENER_CHOICE

Catalog No. IC4728N100
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
IC4728N100 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. IC4728N100 Supplier R&D Systems Supplier No. IC4728N100UG
Add to Cart
Add to Cart
This item is not returnable. View return policy

Mouse Monoclonal Antibody

Neuromedin BR/NMBR Monoclonal specifically detects Neuromedin BR/NMBR in Human samples. It is validated for Intracellular Staining by Flow Cytometry.

Specifications

Antigen Neuromedin BR/NMBR
Applications Flow Cytometry
Classification Monoclonal
Clone 466505
Concentration 0.2 mg/mL
Conjugate Alexa Fluor 700
Dilution Intracellular Staining by Flow Cytometry 0.25-1 ug/10^6 cells
Formulation Supplied 0.2 mg/mL in a saline solution containing BSA and Sodium Azide. with 0.09% Sodium Azide
Gene Accession No. AAB27330
Gene Alias Neuromedin B R, neuromedin B receptor, neuromedin-B receptor, Neuromedin-B-preferring bombesin receptor, NMBR, NMB-R
Gene Symbols NMBR
Host Species Mouse
Immunogen NS0 mouse myeloma cell line transfected with human NMBR Met1-Met390 Accession # AAB27330
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 4829
Test Specificity Detects human Neuromedin B R/NMBR. Stains human Neuromedin B R/NMBR transfectants but not irrelevant transfectants.
Target Species Human
Content And Storage Store the unopened product at 2 - 8 degreesC. Do not use past expiration date.
Isotype IgG2a
Show More Show Less

For Research Use Only (RUO)

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.