Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human Neuromedin BR/NMBR Antibody, R&D Systems™

Mouse Monoclonal Antibody
Supplier: R&D Systems MAB47281SP
This item is not returnable.
View return policy
Description
Neuromedin BR/NMBR Monoclonal specifically detects Neuromedin BR/NMBR in Human samples. It is validated for Immunohistochemistry.Specifications
Neuromedin BR/NMBR | |
Monoclonal | |
LYOPH | |
Immunohistochemistry 8-25 ug/mL | |
NP_002502 | |
NMBR | |
NS0 mouse myeloma cell line transfected with human Neuromedin B R/NMBR Accession # NP_002502 | |
25 μg | |
Primary | |
Detects human Neuromedin B R/NMBR in direct ELISAs. | |
Human | |
Purified |
Immunohistochemistry | |
466501 | |
Unconjugated | |
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
Neuromedin B R, neuromedin B receptor, neuromedin-B receptor, Neuromedin-B-preferring bombesin receptor, NMBR, NMB-R | |
Mouse | |
Protein A or G purified from hybridoma culture supernatant | |
RUO | |
4829 | |
Sterile PBS to a final concentration of 0.5 mg/mL. | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
IgG2b |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction