Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human Neurophysin II Antibody, R&D Systems™

Goat Polyclonal Antibody
$200.50 - $538.00
Specifications
Antigen | Neurophysin II/Arg-vasopressin |
---|---|
Concentration | LYOPH |
Dilution | Western Blot 0.1 μg/mL, Immunohistochemistry 3-15 μg/mL |
Applications | Western Blot, Immunohistochemistry |
Classification | Polyclonal |
Description
Neurophysin II/Arg-vasopressin Polyclonal specifically detects Neurophysin II/Arg-vasopressin in Human samples. It is validated for Western Blot, Immunohistochemistry.Specifications
Neurophysin II/Arg-vasopressin | |
Western Blot 0.1 μg/mL, Immunohistochemistry 3-15 μg/mL | |
Polyclonal | |
Goat | |
Human | |
arginine vasopressin, ARVP, AVP, AVP-NPII, AVRP, neurohypophyseal, NP2, vasopressin-neurophysin 2-copeptin, VP | |
AVP | |
IgG | |
Affinity Purified | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
LYOPH | |
Western Blot, Immunohistochemistry | |
Unconjugated | |
RUO | |
P01185 | |
551 | |
Mouse myeloma cell line NS0-derived recombinant human Neurophysin II Cys20-Tyr164 Accession # P01185 | |
Primary | |
Reconstitute at 0.2 mg/mL in sterile PBS. | |
Detects human Neurophysin II in direct ELISAs and Western blots. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title