Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Novus Biologicals™ Human Neurophysin II/Arg-vasopressin ELISA Kit (Colorimetric)
SDP

Catalog No. NBP262140 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
5 Kit
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP262140 5 Kit
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP262140 Supplier Novus Biologicals™ Supplier No. NBP262140
Add to Cart
Add to Cart

ELISA kits are ideal for the quantification of target antigens in tissue extracts, serum and cell culture.

The Human Neurophysin II/Arg-vasopressin ELISA Kit (Colorimetric) measures human Neurophysin II/Arg-vasopressin in human serum, human plasma, tissue culture media.

Specifications

Assay Range 4.10 - 1,000 pg/ml
Assay Sensitivity 2.84 pg/ml
Product Type ELISA Kit (Colorimetric)
Sample Type Human Plasma, Human Serum, Tissue Culture Media
For Use With (Application) ELISA
Host Species Human
Quantity 5 Kit
Storage Requirements Storage of components varies. See protocol for specific instructions.
Target Arg8-Vasopressin
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.