Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human Neuropilin-1 Alexa Fluor 594-conjugated Antibody, R&D Systems™

Mouse Monoclonal Antibody
Supplier: R&D Systems FAB3870T100UG
This item is not returnable.
View return policy
Description
Neuropilin-1 Monoclonal specifically detects Neuropilin-1 in Human samples. It is validated for Flow Cytometry.Specifications
| Neuropilin-1 | |
| Monoclonal | |
| 0.2 mg/mL | |
| Flow Cytometry 0.25-1 ug/10^6 cells | |
| NP_001019799 | |
| NRP1 | |
| Mouse myeloma cell line NS0-derived recombinant human Neuropilin-1 Phe22-Lys644 Accession # NP_001019799 | |
| RUO | |
| 8829 | |
| Human | |
| IgG2a |
| Flow Cytometry | |
| 446921 | |
| Alexa Fluor 594 | |
| Supplied 0.2 mg/mL in a saline solution containing BSA and Sodium Azide. with 0.09% Sodium Azide | |
| BDCA4, BDCA-4, CD304, CD304 antigen, DKFZp686A03134, DKFZp781F1414, neuropilin 1, neuropilin-1, Neuropilin1, NP1, NRP, NRP1, transmembrane receptor, Vascular endothelial cell growth factor 165 receptor, VEGF165R | |
| Mouse | |
| 100 μg | |
| Primary | |
| Detects human Neuropilin-1 in direct ELISAs. | |
| Store the unopened product at 2 - 8 degreesC. Do not use past expiration date. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only (RUO)
Spot an opportunity for improvement?Share a Content Correction