Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human Neuropilin-2 Antibody, R&D Systems™

Mouse Monoclonal Antibody
Supplier: R&D Systems MAB2215
This item is not returnable.
View return policy
Description
Neuropilin-2 Monoclonal specifically detects Neuropilin-2 in Human samples. It is validated for Western Blot.Specifications
Neuropilin-2 | |
Monoclonal | |
LYOPH | |
Western Blot 1 ug/mL | |
NP_003863 | |
NRP2 | |
Mouse myeloma cell line NS0-derived recombinant human Neuropilin-2 Arg21-Tyr855 Accession # NP_003863 | |
100 μg | |
Primary | |
Detects human Neuropilin-2 in direct ELISAs and Western blots. In direct ELISAs and Western blots, this antibody does not cross-react with recombinant rat (rr) Neuropilin-1 or rrNeuropilin-2. | |
Human | |
Purified |
Western Blot | |
257107 | |
Unconjugated | |
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
MGC126574, neuropilin 2, neuropilin-2, Neuropilin2, neuropilin-2a(22), neuropilin-2b(0), NP2, NPN 2, NPN2, PRO2714, receptor for VEGF165 and semaphorins class3, Vascular endothelial cell growth factor 165 receptor 2, vascular endothelial growth factor-165 receptor 2, VEGF1265R2, VEGF165R2neuropilin-2a(17) | |
Mouse | |
Protein A or G purified from hybridoma culture supernatant | |
RUO | |
8828 | |
Reconstitute at 0.5 mg/mL in sterile PBS. | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
IgG2b |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction