Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Human Neuroplastin Isoform 1 Biotinylated Antibody, R&D Systems™
GREENER_CHOICE

Catalog No. BAF5174
Change view
Click to view available options
Quantity:
50 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BAF5174 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. BAF5174 Supplier R&D Systems Supplier No. BAF5174
Add to Cart
Add to Cart
This item is not returnable. View return policy

Sheep Polyclonal Antibody

Neuroplastin Polyclonal specifically detects Neuroplastin in Human samples. It is validated for Western Blot.

Specifications

Antigen Neuroplastin
Applications Western Blot
Classification Polyclonal
Concentration LYOPH
Conjugate Biotin
Dilution Western Blot 0.1 ug/mL
Gene Accession No. Q9Y639.1
Gene Alias AW554172, GP55, GP55Stromal cell-derived receptor 1, GP65SDR-1, GP66, MGC102805, neuroplastin, np55, Np65, NPTN, SDFR1, SDFR1DKFZp686L2477, SDR1, SDR1stromal cell derived factor receptor 1
Gene Symbols NPTN
Host Species Sheep
Immunogen Mouse myeloma cell line NS0-derived recombinant human Neuroplastin 55 isoform 1 Gln29-Ser219 Accession # Q9Y639.1
Purification Method Affinity Purified
Quantity 50 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 27020
Test Specificity Detects human Neuroplastin in Western blots.
Reconstitution Reconstitute at 0.2 mg/mL in sterile PBS.
Target Species Human
Content And Storage Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.