Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human Neurturin Antibody, R&D Systems™

Goat Polyclonal Antibody has been used in 3 publications
Supplier: R&D Systems AF387SP
This item is not returnable.
View return policy
Description
Neurturin Polyclonal specifically detects Neurturin in Human samples. It is validated for Western Blot, Immunohistochemistry, Neutralization.Specifications
Neurturin | |
Polyclonal | |
Unconjugated | |
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
neurturin, NRTN, NTN | |
Goat | |
Affinity Purified | |
RUO | |
4902 | |
Reconstitute at 0.2 mg/mL in sterile PBS. | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
Western Blot, Immunohistochemistry, Neutralization | |
LYOPH | |
Western Blot 0.1 ug/mL, Immunohistochemistry 5-15 ug/mL, Neutralization | |
Q99748 | |
NRTN | |
E. coli-derived recombinant human Neurturin Ala96-Val197 Accession # Q99748 | |
25 μg | |
Primary | |
Detects human Neurturin in direct ELISAs and Western blots. In direct ELISAs, less than 1% cross-reactivity with recombinant mouse Neurturin is observed. | |
Human | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction