Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Human Neurturin Antibody, R&D Systems™
GREENER_CHOICE

Catalog No. MAB387SP
Change view
Click to view available options
Quantity:
25 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MAB387SP 25 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. MAB387SP Supplier R&D Systems Supplier No. MAB387SP
Add to Cart
Add to Cart
This item is not returnable. View return policy

Mouse Monoclonal Antibody

Neurturin Monoclonal specifically detects Neurturin in Human samples. It is validated for Western Blot, Blockade of Receptor-ligand Interaction.

Specifications

Antigen Neurturin
Applications Western Blot, Blockade of Receptor-ligand Interaction
Classification Monoclonal
Clone 126422
Conjugate Unconjugated
Dilution Western Blot 1 ug/mL, Blockade of Receptor-ligand Interaction
Formulation Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative
Gene Accession No. Q99748
Gene Alias neurturin, NRTN, NTN
Gene Symbols NRTN
Host Species Mouse
Immunogen E. coli-derived recombinant human Neurturin Ala96-Val197 Accession # Q99748
Purification Method Protein A or G purified from hybridoma culture supernatant
Quantity 25 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 4902
Test Specificity Detects human Neurturin in direct ELISAs and Western blots. In direct ELISAs, approximately 80% cross-reactivity with recombinant mouse Neurturin is observed and no cross-reactivity with recombinant mouse (rm) Artemin, recombinant human (rh) Cripto-1, recombinant Drosophila DPP, rhGDNF, rhLefty A, rmLefty-1, rhMIS, rmNodal, rmPersephin, rhTGF-beta 1, rhLatent TGF-beta 1, rhTGF-beta 1.2, rhTGF-beta 2 or rhTGF-beta 3 is observed.
Reconstitution Reconstitute at 0.5 mg/mL in sterile PBS.
Target Species Human
Content And Storage Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Form Purified
Isotype IgG2b
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.