Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human Neurturin Biotinylated Antibody, R&D Systems™

Goat Polyclonal Antibody
Supplier: R&D Systems BAF387
This item is not returnable.
View return policy
Description
Neurturin Polyclonal specifically detects Neurturin in Human samples. It is validated for Western Blot.Specifications
Neurturin | |
Polyclonal | |
Biotin | |
Q99748 | |
NRTN | |
E. coli-derived recombinant human Neurturin Ala96-Val197 Accession # Q99748 | |
50 μg | |
Primary | |
Detects human Neurturin in Western blots. In Western blots, approximately 10% cross-reactivity with recombinant mouse Neurturin is observed. | |
Human | |
IgG |
Western Blot | |
LYOPH | |
Western Blot 0.1 ug/mL | |
neurturin, NRTN, NTN | |
Goat | |
Affinity Purified | |
RUO | |
4902 | |
Reconstitute at 0.2 mg/mL in sterile PBS. | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction