Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Human Neutrophil Elastase/ELA2 Alexa Fluor 594-conjugated Antibody, R&D Systems™
GREENER_CHOICE

Catalog No. IC91671T100
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
IC91671T100 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. IC91671T100 Supplier R&D Systems Supplier No. IC91671T100UG
Add to Cart
Add to Cart
This item is not returnable. View return policy

Mouse Monoclonal Antibody

Neutrophil Elastase/ELA2 Monoclonal specifically detects Neutrophil Elastase/ELA2 in Human samples. It is validated for Intracellular Staining by Flow Cytometry.

Specifications

Antigen Neutrophil Elastase/ELA2
Applications Flow Cytometry
Classification Monoclonal
Clone 950317
Conjugate Alexa Fluor 594
Dilution Intracellular Staining by Flow Cytometry 0.25-1 ug/10^6 cells
Formulation Supplied 0.2 mg/mL in a saline solution containing BSA and Sodium Azide.
Gene Accession No. P08246
Gene Alias Bone marrow serine protease, EC 3.4.21, EC 3.4.21.37, ELA2, ELA2granulocyte-derived elastase, ELANE, elastase 2, neutrophil, elastase, neutrophil expressed, Elastase-2, GE, HLEelastase-2, HNE, Human leukocyte elastase, Leukocyte Elastase, medullasin, NE, neutrophil elastase, PMN elastase, PMN-E, polymorphonuclear elastase, SCN1
Host Species Mouse
Immunogen Chinese hamster ovary cell line CHO-derived recombinant human ELA2 Met1-Asn252 Accession # P08246
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 1991
Test Specificity Detects human ELA2 in direct ELISAs and Western blots.
Target Species Human
Content And Storage Store the unopened product at 2 - 8 degreesC. Do not use past expiration date.
Isotype IgG1
Show More Show Less

For Research Use Only (RUO)

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.