Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human Neutrophil Elastase/ELA2 Alexa Fluor 700-conjugated Antibody, R&D Systems™

Mouse Monoclonal Antibody
Supplier: R&D Systems IC91671RN100UG
This item is not returnable.
View return policy
Description
Neutrophil Elastase/ELA2 Monoclonal specifically detects Neutrophil Elastase/ELA2 in Human samples. It is validated for Intracellular Staining by Flow Cytometry.Specifications
Neutrophil Elastase/ELA2 | |
Monoclonal | |
Alexa Fluor 700 | |
Supplied 0.2 mg/mL in a saline solution containing BSA and Sodium Azide. | |
Bone marrow serine protease, EC 3.4.21, EC 3.4.21.37, ELA2, ELA2granulocyte-derived elastase, ELANE, elastase 2, neutrophil, elastase, neutrophil expressed, Elastase-2, GE, HLEelastase-2, HNE, Human leukocyte elastase, Leukocyte Elastase, medullasin, NE, neutrophil elastase, PMN elastase, PMN-E, polymorphonuclear elastase, SCN1 | |
Chinese hamster ovary cell line CHO-derived recombinant human Neutrophil Elastase/ELA2, Met1-Asn252, Accession # P08246 | |
100 μg | |
Primary | |
Human | |
Purified |
Flow Cytometry | |
950317R | |
Intracellular Staining by Flow Cytometry 0.25-1 ug/10^6 cells | |
P08246 | |
Mouse | |
Protein A or G purified from cell culture supernatant | |
RUO | |
1991 | |
Protect from light. Do not freeze. 12 months from date of receipt, 2 to 8°C as supplied. | |
IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction