Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human Neutrophil Elastase/ELA2 Alexa Fluor 750-conjugated Antibody, R&D Systems™

Mouse Monoclonal Antibody
Supplier: R&D Systems IC91671S100UG
This item is not returnable.
View return policy
Description
Neutrophil Elastase/ELA2 Monoclonal specifically detects Neutrophil Elastase/ELA2 in Human samples. It is validated for Intracellular Staining by Flow Cytometry.Specifications
Neutrophil Elastase/ELA2 | |
Monoclonal | |
Alexa Fluor 750 | |
Supplied 0.2 mg/mL in a saline solution containing BSA and Sodium Azide. | |
Bone marrow serine protease, EC 3.4.21, EC 3.4.21.37, ELA2, ELA2granulocyte-derived elastase, ELANE, elastase 2, neutrophil, elastase, neutrophil expressed, Elastase-2, GE, HLEelastase-2, HNE, Human leukocyte elastase, Leukocyte Elastase, medullasin, NE, neutrophil elastase, PMN elastase, PMN-E, polymorphonuclear elastase, SCN1 | |
Chinese hamster ovary cell line CHO-derived recombinant human ELA2 Met1-Asn252 Accession # P08246 | |
RUO | |
1991 | |
Human | |
IgG1 |
Flow Cytometry | |
950317 | |
Intracellular Staining by Flow Cytometry 0.25-1 ug/10^6 cells | |
P08246 | |
Mouse | |
100 μg | |
Primary | |
Detects human ELA2 in direct ELISAs and Western blots. | |
Store the unopened product at 2 - 8 degreesC. Do not use past expiration date. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only (RUO)
Spot an opportunity for improvement?Share a Content Correction