Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human Neutrophil Elastase/ELA2 Antibody, R&D Systems™

Mouse Monoclonal Antibody has been used in 2 publications
$227.50 - $584.50
Specifications
Antigen | Neutrophil Elastase/ELA2 |
---|---|
Clone | 950317 |
Dilution | Western Blot 0.1 ug/mL, Immunohistochemistry 3-25 ug/mL, Intracellular Staining by Flow Cytometry 0.25 ug/10^6 cells, Immunocytochemistry 5-25 ug/mL, CyTOF-ready |
Applications | Western Blot, Immunohistochemistry, Flow Cytometry, Immunocytochemistry, CyTOF |
Classification | Monoclonal |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
MAB91671SP
![]() |
R&D Systems
MAB91671SP |
25 μg |
Each for $227.50
|
|
|||||
MAB91671100
![]() |
R&D Systems
MAB91671100 |
100 μg |
Each for $584.50
|
|
|||||
Description
Neutrophil Elastase/ELA2 Monoclonal specifically detects Neutrophil Elastase/ELA2 in Human samples. It is validated for Western Blot, Immunohistochemistry, Intracellular Staining by Flow Cytometry, Immunocytochemistry, CyTOF-ready.Specifications
Neutrophil Elastase/ELA2 | |
Western Blot 0.1 ug/mL, Immunohistochemistry 3-25 ug/mL, Intracellular Staining by Flow Cytometry 0.25 ug/10^6 cells, Immunocytochemistry 5-25 ug/mL, CyTOF-ready | |
Monoclonal | |
Purified | |
RUO | |
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
Bone marrow serine protease, EC 3.4.21, EC 3.4.21.37, ELA2, ELA2granulocyte-derived elastase, ELANE, elastase 2, neutrophil, elastase, neutrophil expressed, Elastase-2, GE, HLEelastase-2, HNE, Human leukocyte elastase, Leukocyte Elastase, medullasin, NE, neutrophil elastase, PMN elastase, PMN-E, polymorphonuclear elastase, SCN1 | |
Chinese hamster ovary cell line CHO-derived recombinant human ELA2 Met1-Asn252 Accession # P08246 | |
Primary | |
Reconstitute at 0.5 mg/mL in sterile PBS. | |
Detects human ELA2 in direct ELISAs and Western blots. |
950317 | |
Western Blot, Immunohistochemistry, Flow Cytometry, Immunocytochemistry, CyTOF | |
Unconjugated | |
Mouse | |
Human | |
P08246 | |
1991 | |
IgG1 | |
Protein A or G purified from hybridoma culture supernatant | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title