Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Novus Biologicals™ Human NF-H [p Ser668, p Ser702] ELISA Kit (Colorimetric)
SDP

Catalog No. NB001549 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
1 Kit
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB001549 1 Kit
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB001549 Supplier Novus Biologicals™ Supplier No. NBP278763
Add to Cart
Add to Cart

Specifications

Assay Range 78.13 to 5000 pg/mL
Assay Sensitivity 46.88 pg/mL
Product Type ELISA Kit (Colorimetric)
Sample Type Other Biological Fluids, Plasma, Serum
Specificity This kit recognizes Human pNF-H in samples. No significant cross-reactivity or interference between Human pNF-H and analogues was observed
For Use With (Application) ELISA
Gene ID (Entrez) 4744
Gene Symbol NEFH
Host Species Human
Quantity 1 Kit
Research Discipline Autophagy, Cell Biology, Cellular Markers, Neurodegeneration, Neuronal Cell Markers, Neuroscience
Storage Requirements Storage of components varies. See protocol for specific instructions.
Target NF-H
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.