Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human NF-L Antibody, R&D Systems™

Mouse Monoclonal Antibody
$200.50 - $491.00
Specifications
Antigen | NF-L |
---|---|
Clone | 1033955 |
Dilution | Western Blot 2 ug/mL, ELISA, Immunohistochemistry 5-25 ug/mL |
Applications | Western Blot, ELISA, Immunohistochemistry |
Classification | Monoclonal |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
MAB22164SP
![]() |
R&D Systems
MAB22164SP |
25 μg |
Each for $200.50
|
|
|||||
MAB22164100
![]() |
R&D Systems
MAB22164100 |
100 μg |
Each for $491.00
|
|
|||||
Description
NF-L Monoclonal antibody specifically detects NF-L in Human samples. It is validated for Western Blot, ELISA, ImmunohistochemistrySpecifications
NF-L | |
Western Blot 2 ug/mL, ELISA, Immunohistochemistry 5-25 ug/mL | |
Monoclonal | |
Purified | |
RUO | |
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. | |
4747 | |
IgG2b | |
Protein A or G purified from hybridoma culture supernatant | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70°C as supplied. 1 month, 2 to 8°C under sterile conditions after reconstitution. 6 months, -20 to -70°C under sterile conditions after reconstitution. |
1033955 | |
Western Blot, ELISA, Immunohistochemistry | |
Unconjugated | |
Mouse | |
Human | |
68 kDa neurofilament protein, CMT2E, NEFL, neurofilament protein, light chain, neurofilament subunit NF-L, Neurofilament triplet L protein, neurofilament, light polypeptide, neurofilament-light, NF68, NF68FLJ53642, NFL, NF-L, NFLlight polypeptide 68kDa | |
Human NF-L peptide | |
Primary | |
Reconstitute at 0.5 mg/mL in sterile PBS. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title