Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human NF-L Antibody, R&D Systems™

Mouse Monoclonal Antibody
$200.50 - $467.50
Specifications
| Antigen | NF-L |
|---|---|
| Clone | 1033926 |
| Dilution | ELISA |
| Applications | ELISA |
| Classification | Monoclonal |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
MAB22165SP
![]() |
R&D Systems
MAB22165SP |
25 μg |
Each for $200.50
|
|
|||||
MAB22165100
![]() |
R&D Systems
MAB22165100 |
100 μg |
Each for $467.50
|
|
|||||
Description
NF-L Monoclonal antibody specifically detects NF-L in Human samples. It is validated for ELISASpecifications
| NF-L | |
| ELISA | |
| Monoclonal | |
| Purified | |
| RUO | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. | |
| 4747 | |
| IgG2b | |
| Protein A or G purified from hybridoma culture supernatant | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70°C as supplied. 1 month, 2 to 8°C under sterile conditions after reconstitution. 6 months, -20 to -70°C under sterile conditions after reconstitution. |
| 1033926 | |
| ELISA | |
| Unconjugated | |
| Mouse | |
| Human | |
| 68 kDa neurofilament protein, CMT2E, NEFL, neurofilament protein, light chain, neurofilament subunit NF-L, Neurofilament triplet L protein, neurofilament, light polypeptide, neurofilament-light, NF68, NF68FLJ53642, NFL, NF-L, NFLlight polypeptide 68kDa | |
| Human NF-L synthetic peptide | |
| Primary | |
| Reconstitute at 0.5 mg/mL in sterile PBS. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title