Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Human NF-L Antibody, R&D Systems™
GREENER_CHOICE

Catalog No. MAB22162100
Change view
Click to view available options
Quantity:
100 μg
25 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MAB22162100 100 μg
MAB22162SP 25 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. MAB22162100 Supplier R&D Systems Supplier No. MAB22162100
Add to Cart
Add to Cart
This item is not returnable. View return policy

Mouse Monoclonal Antibody

NF-L Monoclonal specifically detects NF-L in Human samples. It is validated for Immunocytochemistry.

Specifications

Antigen NF-L
Applications Immunocytochemistry
Classification Monoclonal
Clone 985917
Conjugate Unconjugated
Dilution Immunocytochemistry 8-25μg/mL
Formulation Lyophilized from a 0.2μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2μm filtered solution in PBS.
Gene Accession No. P07196
Gene Alias 68 kDa neurofilament protein, CMT2E, NEFL, neurofilament protein, light chain, neurofilament subunit NF-L, Neurofilament triplet L protein, neurofilament, light polypeptide, neurofilament-light, NF68, NF68FLJ53642, NFL, NF-L, NFLlight polypeptide 68kDa
Host Species Mouse
Immunogen E. coli-derived recombinant human NF-L Met1-Asp338 Accession # P07196
Purification Method Protein A or G purified from hybridoma culture supernatant
Quantity 100 μg
Primary or Secondary Primary
Gene ID (Entrez) 4747
Test Specificity Detects human NF-L in direct ELISAs.
Reconstitution Reconstitute at 0.5mg/mL in sterile PBS.
Target Species Human
Content And Storage Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Form Supernatant
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.