Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Human NF-M Antibody, R&D Systems™
GREENER_CHOICE

Catalog No. AF3029SP
Change view
Click to view available options
Quantity:
100 μg
25 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AF3029SP 25 μg
AF3029 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. AF3029SP Supplier R&D Systems Supplier No. AF3029SP
Add to Cart
Add to Cart
This item is not returnable. View return policy

Sheep Polyclonal Antibody

NF-M Polyclonal specifically detects NF-M in Human samples. It is validated for Western Blot, Immunohistochemistry, Knockout Validated.

Specifications

Antigen NF-M
Applications Western Blot, Immunohistochemistry, KnockDown
Classification Polyclonal
Concentration LYOPH
Conjugate Unconjugated
Dilution Western Blot 1-2 ug/mL, Immunohistochemistry 5-15 ug/mL, Knockout Validated
Formulation Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative
Gene Accession No. P07197
Gene Alias NEF3, NEF3neurofilament medium polypeptide, NEFM, Neurofilament 3, neurofilament, medium polypeptide, neurofilament, medium polypeptide 150kDa, neurofilament-3 (150 kD medium), NFM, NF-M160 kDa neurofilament protein, NFMNeurofilament triplet M protein
Gene Symbols NEFM
Host Species Sheep
Immunogen E. coli-derived recombinant human NF-M Ser2-Ser358 Accession # P07197
Purification Method Affinity Purified
Quantity 25 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 4741
Test Specificity Detects human NF-M in direct ELISAs and Western blots. In direct ELISAs and Western blots, approximately 5% cross-reactivity with recombinant human (rh) NF-H and rhNF-L is observed.
Reconstitution Reconstitute at 0.2 mg/mL in sterile PBS.
Target Species Human
Content And Storage Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.