Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Human NF2/Merlin Antibody, R&D Systems™
GREENER_CHOICE

Catalog No. MAB5616
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MAB5616 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. MAB5616 Supplier R&D Systems Supplier No. MAB5616
Add to Cart
Add to Cart
This item is not returnable. View return policy

Mouse Monoclonal Antibody

NF2/Merlin Monoclonal specifically detects NF2/Merlin in Human samples. It is validated for Western Blot.

Specifications

Antigen NF2/Merlin
Applications Western Blot
Classification Monoclonal
Clone 534428
Concentration LYOPH
Conjugate Unconjugated
Dilution Western Blot 2 ug/mL
Formulation Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative
Gene Accession No. P35240
Gene Alias ACN, BANF, Merlin, moesin-ezrin-radixin like, Moesin-ezrin-radixin-like protein, moesin-ezrin-radizin-like protein, neurofibromin 2 (bilateral acoustic neuroma), neurofibromin 2 (merlin), neurofibromin-2, SCH, Schwannomerlin, Schwannomin
Gene Symbols NF2
Host Species Mouse
Immunogen E. coli-derived recombinant human NF2/Merlin Met363-Lys578 Accession # P35240
Purification Method Protein A or G purified from hybridoma culture supernatant
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 4771
Test Specificity Detects human NF2/Merlin in direct ELISAs and Western blots. In direct ELISAs, no cross-reactivity with recombinant human NF1 is observed.
Reconstitution Reconstitute at 0.5 mg/mL in sterile PBS.
Target Species Human
Content And Storage Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Form Purified
Isotype IgG2b
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.