Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Human NFIL3/E4BP4 Alexa Fluor 594-conjugated Antibody, R&D Systems™
GREENER_CHOICE

Catalog No. FAB8570T100
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
FAB8570T100 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. FAB8570T100 Supplier R&D Systems Supplier No. FAB8570T100UG
Add to Cart
Add to Cart
This item is not returnable. View return policy

Mouse Monoclonal Antibody

NFIL3 Monoclonal specifically detects NFIL3 in Human samples. It is validated for Flow Cytometry.

Specifications

Antigen NFIL3
Applications Flow Cytometry
Classification Monoclonal
Clone 714401
Concentration 0.2 mg/mL
Conjugate Alexa Fluor 594
Dilution Flow Cytometry 0.25-1 ug/10^6 cells
Formulation Supplied 0.2 mg/mL in a saline solution containing BSA and Sodium Azide. with 0.09% Sodium Azide
Gene Accession No. Q16649
Gene Alias E4BP4, E4BP4IL3BP1, IL3BP1, Interleukin-3 promoter transcriptional activator, Interleukin-3-binding protein 1, NF-IL3A, NFIL3AE4 promoter-binding protein 4, nuclear factor interleukin 3 regulated protein, nuclear factor interleukin-3-regulated protein, nuclear factor, interleukin 3 regulated, Transcriptional activator NF-IL3A
Gene Symbols NFIL3
Host Species Mouse
Immunogen E. coli-derived recombinant human NFIL3/E4BP4 Lys140-Gly462 Accession # Q16649
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 4783
Test Specificity Detects human NFIL3/E4BP4 in direct ELISAs.
Target Species Human
Content And Storage Store the unopened product at 2 - 8 degreesC. Do not use past expiration date.
Isotype IgG1
Show More Show Less

For Research Use Only (RUO)

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.