Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Human NFkB2 Antibody, R&D Systems™
GREENER_CHOICE

Catalog No. MAB28881SP
Change view
Click to view available options
Quantity:
25 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MAB28881SP 25 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. MAB28881SP Supplier R&D Systems Supplier No. MAB28881SP
Add to Cart
Add to Cart
This item is not returnable. View return policy

Mouse Monoclonal Antibody

NFkB2/NFkB p100 Monoclonal specifically detects NFkB2/NFkB p100 in Human samples. It is validated for Western Blot, Chromatin Immunoprecipitation (ChIP), Immunocytochemistry.

Specifications

Antigen NFkB2/NFkB p100
Applications ChIP Assay, Immunocytochemistry
Classification Monoclonal
Clone 291319
Concentration LYOPH
Conjugate Unconjugated
Dilution Western Blot 0.5 ug/mL, Chromatin Immunoprecipitation (ChIP) 5 ug/5 x 10^6 cells, Immunocytochemistry 3-25 ug/mL
Formulation Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative
Gene Accession No. Q00653
Gene Symbols NFKB2
Host Species Mouse
Immunogen E. coli-derived recombinant human NF kappa B2 Met1-Asn447 Accession # Q00653
Purification Method Protein A or G purified from hybridoma culture supernatant
Quantity 25 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 4791
Test Specificity Detects human NFkB2 in direct ELISAs and Western blots. In Western blots, no cross-reactivity with endogenous mouse NFkB2 in NIH 3T3 cells is observed.
Reconstitution Reconstitute at 0.5 mg/mL in sterile PBS.
Target Species Human
Content And Storage Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Form Purified
Isotype IgG1
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.