Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human NG2/MCSP Antibody, R&D Systems™

Goat Polyclonal Antibody
Supplier: R&D Systems AF2585
This item is not returnable.
View return policy
Description
NG2/MCSP Polyclonal specifically detects NG2/MCSP in Human samples. It is validated for Western Blot, Knockout Validated.Specifications
| NG2/MCSP | |
| Polyclonal | |
| Unconjugated | |
| Q6UVK1 | |
| CSPG4 | |
| Mouse myeloma cell line NS0-derived recombinant human NG2/MCSP Ser1583-Ser2224 Accession # Q6UVK1 | |
| 100 μg | |
| Primary | |
| Detects human NG2/MCSP in direct ELISAs and Western blots. In Western blots, approximately 30% cross-reactivity with recombinant mouse CSPG4 is observed. | |
| Human | |
| IgG |
| Western Blot | |
| LYOPH | |
| Western Blot 0.5 ug/mL | |
| AN2, chondroitin sulfate proteoglycan 4, chondroitin sulfate proteoglycan 4 (melanoma-associated), CSPG4, EC 3.6.3, HMW-MAA, MCSP, MCSPChondroitin sulfate proteoglycan NG2, MCSPG, MCSPGMelanoma chondroitin sulfate proteoglycan, MEL-CSPG, MSK16Melanoma-associated chondroitin sulfate proteoglycan, NG2EC 2.7.8 | |
| Goat | |
| Affinity Purified | |
| RUO | |
| 1464 | |
| Reconstitute at 0.2 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction