Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Human NGFI-B gamma/NOR-1/NR4A3 Antibody, R&D Systems™
GREENER_CHOICE

Catalog No. PPH783300
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PPH783300 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. PPH783300 Supplier R&D Systems Supplier No. PPH783300
Add to Cart
Add to Cart
This item is not returnable. View return policy

Mouse Monoclonal Antibody has been used in 9 publications

NGFI-B gamma/NOR-1/NR4A3 Monoclonal specifically detects NGFI-B gamma/NOR-1/NR4A3 in Human samples. It is validated for Western Blot, Immunoprecipitation.

Specifications

Antigen NGFI-B gamma/NOR-1/NR4A3
Applications Western Blot, Immunoprecipitation
Classification Monoclonal
Clone H7833
Conjugate Unconjugated
Dilution Western Blot, Immunoprecipitation
Formulation A liquid formulation in physiologic saline with 0.1% NaN 3 . with 0.1% Sodium Azide
Gene Symbols NR4A3
Host Species Mouse
Immunogen Recombinant human NGFI-Bγ (amino acids 2 - 99) and mouse myeloma cells (NS-1)
Purification Method Protein A or G purified from hybridoma culture supernatant
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 8013
Target Species Human
Content And Storage Store the unopened product at -70 degreesC. Use a manual defrost freezer and avoid repeated freeze-thaw cycles. Do not use past expiration date.
Form Purified
Isotype IgG2a
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.