Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Human NGFR/TNFRSF16 Antibody, R&D Systems™
GREENER_CHOICE

Catalog No. AF367
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AF367 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. AF367 Supplier R&D Systems Supplier No. AF367
Add to Cart
Add to Cart
This item is not returnable. View return policy

Goat Polyclonal Antibody

NGFR/TNFRSF16/p75NTR Polyclonal specifically detects NGFR/TNFRSF16/p75NTR in Human samples. It is validated for ELISA Capture (Matched Antibody Pair).

Specifications

Antigen NGFR/TNFRSF16/p75NTR
Applications ELISA
Classification Polyclonal
Concentration LYOPH
Conjugate Unconjugated
Dilution ELISA Capture (Matched Antibody Pair) 0.2-0.8 ug/mL
Gene Accession No. P08138
Gene Alias CD271, CD271 antigen, Gp80-LNGFR, member 16, nerve growth factor receptor, NGF R, NGF receptor, p75 ICD, p75 NTR, TNFRSF16, TNFRSF16nerve growth factor receptor (TNFR superfamily, member 16), tumor necrosis factor receptor superfamily member 16
Gene Symbols NGFR
Host Species Goat
Immunogen S. frugiperda insect ovarian cell line Sf 21-derived recombinant human NGF R/TNFRSF16 Lys29-Asn250 Accession # P08138
Purification Method Affinity Purified
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 4804
Test Specificity Detects human NGF R/TNFRSF16 in direct ELISAs. In direct ELISAs, approximately 15% cross-reactivity with recombinant mouse NGF R is observed.
Reconstitution Reconstitute at 0.2 mg/mL in sterile PBS.
Target Species Human
Content And Storage Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.