Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human NGL-1/LRRC4C Antibody, R&D Systems™

Mouse Monoclonal Antibody
Supplier: R&D Systems MAB4899SP
This item is not returnable.
View return policy
Description
NGL-1/LRRC4C Monoclonal specifically detects NGL-1/LRRC4C in Human samples. It is validated for Immunohistochemistry.Specifications
NGL-1/LRRC4C | |
Monoclonal | |
LYOPH | |
Immunohistochemistry 8-25 ug/mL | |
Q9HCJ2 | |
LRRC4C | |
Mouse myeloma cell line NS0-derived recombinant human NGL-1/LRRC4C Gln45-Lys527 (predicted) Accession # Q9HCJ2 | |
25 μg | |
Primary | |
Detects human NGL-1/LRRC4C in direct ELISAs. In direct ELISAs, no cross-reactivity with recombinant human (rh) LRRC4 or rhNGL-3/LRRC4B is observed. | |
Human | |
Purified |
Immunohistochemistry | |
755720 | |
Unconjugated | |
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
KIAA1580NGL-1NGL1Netrin-G1 ligand, leucine rich repeat containing 4C, leucine-rich repeat-containing protein 4C, LRRC4C, NGL1 | |
Mouse | |
Protein A or G purified from hybridoma culture supernatant | |
RUO | |
57689 | |
Sterile PBS to a final concentration of 0.5 mg/mL. | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction