Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Human NGL-1/LRRC4C Biotinylated Antibody, R&D Systems™
GREENER_CHOICE

Catalog No. BAF4899
Change view
Click to view available options
Quantity:
50 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BAF4899 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. BAF4899 Supplier R&D Systems Supplier No. BAF4899
Add to Cart
Add to Cart
This item is not returnable. View return policy

Sheep Polyclonal Antibody

NGL-1/LRRC4C Polyclonal specifically detects NGL-1/LRRC4C in Human samples. It is validated for Western Blot.

Specifications

Antigen NGL-1/LRRC4C
Applications Western Blot
Classification Polyclonal
Concentration LYOPH
Conjugate Biotin
Dilution Western Blot 0.1 ug/mL
Gene Accession No. Q9HCJ2
Gene Alias KIAA1580NGL-1NGL1Netrin-G1 ligand, leucine rich repeat containing 4C, leucine-rich repeat-containing protein 4C, LRRC4C, NGL1
Gene Symbols LRRC4C
Host Species Sheep
Immunogen Mouse myeloma cell line NS0-derived recombinant human NGL-1/LRRC4C Gln45-Lys527 Accession # Q9HCJ2
Purification Method Affinity Purified
Quantity 50 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 57689
Test Specificity Detects human NGL-1/LRRC4C in Western blots.
Reconstitution Reconstitute at 0.2 mg/mL in sterile PBS.
Target Species Human
Content And Storage Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.