Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Human NGL-3/LRRC4B Antibody, R&D Systems™
GREENER_CHOICE

Catalog No. MAB69191
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MAB69191 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. MAB69191 Supplier R&D Systems Supplier No. MAB69191
Add to Cart
Add to Cart
This item is not returnable. View return policy

Mouse Monoclonal Antibody

NGL-3/LRRC4B/LRCH4B Monoclonal specifically detects NGL-3/LRRC4B/LRCH4B in Human samples. It is validated for Western Blot.

Specifications

Antigen NGL-3/LRRC4B/LRCH4B
Applications Western Blot
Classification Monoclonal
Clone 702008
Concentration LYOPH
Conjugate Unconjugated
Dilution Western Blot 1 ug/mL
Formulation Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative
Gene Accession No. Q9NT99
Gene Alias DKFZp761A179, HSM, Leucine Rich Repeat Containing 4B, Leucine-Rich Repeat-Containing Protein 4B, Leucine-Rich Repeats And Immunoglobulin-Like Domains 4, LRIG4, LRRC4B, Netrin-G3 Ligand, NGL3, NGL-3
Gene Symbols LRRC4B
Host Species Mouse
Immunogen Chinese hamster ovary cell line CHO-derived recombinant human NGL-3/LRRC4B Ala47-Lys576 Accession # Q9NT99
Purification Method Protein A or G purified from hybridoma culture supernatant
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 94030
Test Specificity Detects human NGL-3/LRRC4B in direct ELISAs and Western blots. In direct ELISAs and Western blots, no cross-reactivity with recombinant human (rh) LRRC3, rhLRRC4, or rhLRRC32 is observed.
Reconstitution Sterile PBS to a final concentration of 0.5 mg/mL.
Target Species Human
Content And Storage Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Form Purified
Isotype IgG2a
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.