Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Human NgR2/NgRH1 Antibody, R&D Systems™
GREENER_CHOICE

Catalog No. MAB2776SP
Change view
Click to view available options
Quantity:
100 μg
25 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MAB2776SP 25 μg
MAB2776 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. MAB2776SP Supplier R&D Systems Supplier No. MAB2776SP
Add to Cart
Add to Cart
This item is not returnable. View return policy

Mouse Monoclonal Antibody

NgR2/NgRH1/RTN4RL2 Monoclonal specifically detects NgR2/NgRH1/RTN4RL2 in Human samples. It is validated for Western Blot.

Specifications

Antigen NgR2/NgRH1/RTN4RL2
Applications Western Blot
Classification Monoclonal
Clone 333413
Concentration LYOPH
Conjugate Unconjugated
Dilution Western Blot 1 ug/mL
Formulation Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative
Gene Accession No. Q86UN3
Gene Alias NgR2, NGRH1, reticulon 4 receptor-like 2, RTN4RL2
Gene Symbols RTN4RL2
Host Species Mouse
Immunogen Mouse myeloma cell line NS0-derived recombinant human NgR2/NgRH1 Ala28-Leu401 Accession # Q86UN3
Purification Method Protein A or G purified from hybridoma culture supernatant
Quantity 25 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 349667
Test Specificity Detects human NgR2/NgRH1 in direct ELISAs and Western blots. In direct ELISAs and Western blots, no cross-reactivity with recombinant human (rh) NgR3 or rhNgR is observed.
Reconstitution Reconstitute at 0.5 mg/mL in sterile PBS.
Target Species Human
Content And Storage Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Form Purified
Isotype IgG2b
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.