Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Human NHA2/SLC9B2 Alexa Fluor 647-conjugated Antibody, R&D Systems™
GREENER_CHOICE

Catalog No. FAB8404R100
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
FAB8404R100 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. FAB8404R100 Supplier R&D Systems Supplier No. FAB8404R100UG
Add to Cart
Add to Cart
This item is not returnable. View return policy

Mouse Monoclonal Antibody

NHA2/SLC9B2/NHEDC2 Monoclonal specifically detects NHA2/SLC9B2/NHEDC2 in Human samples. It is validated for Flow Cytometry.

Specifications

Antigen NHA2/SLC9B2/NHEDC2
Applications Flow Cytometry
Classification Monoclonal
Clone 896151
Concentration 0.2 mg/mL
Conjugate Alexa Fluor 647
Dilution Flow Cytometry 0.25-1 ug/10^6 cells
Formulation Supplied 0.2 mg/mL in a saline solution containing BSA and Sodium Azide. with 0.09% Sodium Azide
Gene Accession No. NP_849155
Gene Alias FLJ23984, Mitochondrial Na(+)/H(+) exchanger NHA2, mitochondrial sodium/hydrogen exchanger NHA2, Na(+)/H(+) exchanger-like domain-containing protein 2, Na+/H+ exchanger domain containing 2, NHA2NHE10, NHE domain-containing protein 2, NHE10, NHEDC2, SLC9B2, Sodium/hydrogen exchanger-like domain-containing protein 2
Gene Symbols SLC9B2
Host Species Mouse
Immunogen NS0 mouse myeloma cell line transfected with human NHA2/SLC9B2 Met1-Val537 Accession # NP_849155
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 133308
Test Specificity Detects HEK293 cells transfected with human NHA2/SLC9B2 by Flow Cytometry. Does not detect untransfected or irrelevant transfected HEK293 cells.
Target Species Human
Content And Storage Store the unopened product at 2 - 8 degreesC. Do not use past expiration date.
Isotype IgG2b
Show More Show Less

For Research Use Only (RUO)

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.