Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human Nicastrin Antibody, R&D Systems™

Mouse Monoclonal Antibody
Supplier: R&D Systems MAB53781SP
This item is not returnable.
View return policy
Description
Nicastrin Monoclonal specifically detects Nicastrin in Human samples. It is validated for Western Blot.Specifications
Nicastrin | |
Monoclonal | |
LYOPH | |
Western Blot 1 ug/mL | |
Q92542 | |
NCSTN | |
Chinese hamster ovary cell line CHO-derived recombinant human Nicastrin Asn34-Glu669 Accession # Q92542 | |
25 μg | |
Primary | |
Detects human Nicastrin in direct ELISAs and Western blots. | |
Human | |
Purified |
Western Blot | |
716910 | |
Unconjugated | |
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
anterior pharynx-defective 2, APH2, ATAG1874, KIAA0253APH2, NCSTN, nicastrin | |
Mouse | |
Protein A or G purified from hybridoma culture supernatant | |
RUO | |
23385 | |
Sterile PBS to a final concentration of 0.5 mg/mL. | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction