Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human Nicastrin Antibody, R&D Systems™

Mouse Monoclonal Antibody
Supplier: R&D Systems MAB53781
This item is not returnable.
View return policy
Description
Nicastrin Monoclonal specifically detects Nicastrin in Human samples. It is validated for Western Blot.Specifications
Nicastrin | |
Monoclonal | |
LYOPH | |
Western Blot 1 ug/mL | |
Q92542 | |
NCSTN | |
Chinese hamster ovary cell line CHO-derived recombinant human Nicastrin Asn34-Glu669 Accession # Q92542 | |
100 μg | |
Primary | |
Detects human Nicastrin in direct ELISAs and Western blots. | |
Human | |
Purified |
Western Blot | |
716910 | |
Unconjugated | |
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
anterior pharynx-defective 2, APH2, ATAG1874, KIAA0253APH2, NCSTN, nicastrin | |
Mouse | |
Protein A or G purified from hybridoma culture supernatant | |
RUO | |
23385 | |
Sterile PBS to a final concentration of 0.5 mg/mL. | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction