Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Human Nidogen-1/Entactin Antibody, R&D Systems™
GREENER_CHOICE

Catalog No. MAB2570500
Change view
Click to view available options
Quantity:
100 μg
25 μg
500 μg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MAB2570500 500 μg
MAB2570SP 25 μg
MAB2570100 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Catalog No. MAB2570500 Supplier R&D Systems Supplier No. MAB2570500
Add to Cart
Add to Cart
This item is not returnable. View return policy

Mouse Monoclonal Antibody has been used in 3 publications

Nidogen-1/Entactin Monoclonal specifically detects Nidogen-1/Entactin in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry, ELISA Capture (Matched Antibody Pair).

Specifications

Antigen Nidogen-1/Entactin
Applications Western Blot, Immunohistochemistry, Immunocytochemistry, ELISA
Classification Monoclonal
Clone 302117
Conjugate Unconjugated
Dilution Western Blot 1 ug/mL, Immunohistochemistry 8-25 ug/mL, Immunocytochemistry 8-25 ug/mL, ELISA Capture (Matched Antibody Pair) 2-8 ug/mL
Formulation Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative
Gene Accession No. AAH45606.1
Gene Alias enactin, Entactin, Entactin-1, NID1, NID-1, NIDentactin, nidogen (enactin), nidogen 1, nidogen-1, Nidogen1
Gene Symbols NID1
Host Species Mouse
Immunogen Mouse myeloma cell line NS0-derived recombinant human Nidogen-1/Entactin Leu29-Lys1114 (Gln1113Arg) Accession # AAH45606.1
Purification Method Protein A or G purified from hybridoma culture supernatant
Quantity 500 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 4811
Test Specificity Detects human Nidogen-1/Entactin in ELISAs and Western blots. In direct ELISAs and Western blots, no cross-reactivity with recombinant human Nidogen-2 is observed.
Reconstitution Reconstitute at 0.5 mg/mL in sterile PBS.
Target Species Human
Content And Storage Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Form Purified
Isotype IgG1
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.