Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Human Nidogen-2 Biotinylated Antibody, R&D Systems™
GREENER_CHOICE

Catalog No. BAF3385
Change view
Click to view available options
Quantity:
50 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BAF3385 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. BAF3385 Supplier R&D Systems Supplier No. BAF3385
Add to Cart
Add to Cart
This item is not returnable. View return policy

Goat Polyclonal Antibody

Nidogen-2 Polyclonal specifically detects Nidogen-2 in Human samples. It is validated for Western Blot, Immunohistochemistry.

Specifications

Antigen Nidogen-2
Applications Western Blot, Immunohistochemistry
Classification Polyclonal
Concentration LYOPH
Conjugate Biotin
Dilution Western Blot 0.1 ug/mL, Immunohistochemistry 5-15 ug/mL
Gene Accession No. Q14112
Gene Alias NID2, NID-2, nidogen 2 (osteonidogen), nidogen-2, Nidogen2, osteonidogen
Gene Symbols NID2
Host Species Goat
Immunogen Chinese hamster ovary cell line CHO-derived recombinant human Nidogen-2 Leu31-Lys1375 (Gly832Ala) Accession # Q14112
Purification Method Affinity Purified
Quantity 50 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 22795
Test Specificity Detects human Nidogen-2 in Western blots. In Western blots, less than 1% cross-reactivity with recombinant human Nidogen-1 is observed.
Reconstitution Reconstitute at 0.2 mg/mL in sterile PBS.
Target Species Human
Content And Storage Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.