Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Human NIK/MAP3K14 Antibody, R&D Systems™
GREENER_CHOICE

Catalog No. MAB6888RSP
Change view
Click to view available options
Quantity:
100 μg
25 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MAB6888RSP 25 μg
MAB6888R100 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. MAB6888RSP Supplier R&D Systems Supplier No. MAB6888RSP
Add to Cart
Add to Cart
This item is not returnable. View return policy

Mouse Monoclonal Antibody

NIK/MAP3K14 Monoclonal antibody specifically detects NIK/MAP3K14 in Human samples. It is validated for Immunohistochemistry

Specifications

Antigen NIK/MAP3K14
Applications Immunohistochemistry
Classification Monoclonal
Clone 603917R
Conjugate Unconjugated
Dilution Immunohistochemistry 3-25 ug/mL
Formulation Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. Small pack size (SP) is supplied either lyophilized or as a 0.2 μm filtered solution in PBS.
Gene Accession No. Q99558
Gene Alias EC 2.7.11.25, FTDCR1B, HS, HsNIK, HSNIKserine/threonine protein-kinase, MAP3K14, mitogen-activated protein kinase kinase kinase 14, NIKNF-kappa-beta-inducing kinase, Serine/threonine-protein kinase NIK
Host Species Mouse
Immunogen E. coli-derived recombinant human NIK/MAP3K14, Glu769-Pro947, Accession # Q99558
Purification Method Protein A or G purified from hybridoma culture supernatant
Quantity 25 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 9020
Reconstitution Reconstitute at 0.5 mg/mL in sterile PBS.
Target Species Human
Content And Storage Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 °C as supplied. 1 month, 2 to 8 °C under sterile conditions after reconstitution. 6 months, -20 to -70 °C under sterile conditions after reconstitution.
Form Purified
Isotype IgG2a
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.