Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human Ninjurin-1 Alexa Fluor 488-conjugated Antibody, R&D Systems™

Mouse Monoclonal Antibody
Supplier: R&D Systems FAB51051G100UG
This item is not returnable.
View return policy
Description
Ninjurin-1 Monoclonal specifically detects Ninjurin-1 in Human samples. It is validated for Flow Cytometry.Specifications
Ninjurin-1 | |
Monoclonal | |
Alexa Fluor 488 | |
Supplied 0.2 mg/mL in a saline solution containing BSA and Sodium Azide. with Sodium Azide | |
Nerve injury-induced protein 1, nerve injury-induced protein-1, NIN1, NINJ1, NINJURIN, ninjurin 1, ninjurin-1, Ninjurin1 | |
Mouse | |
Protein A or G purified from hybridoma culture supernatant | |
Primary | |
Detects human Ninjurin-1 in direct ELISAs. In direct ELISAs, no cross-reactivity with recombinant human Ninjurin-2 is observed. | |
Protect from light. Do not freeze. 12 months from date of receipt, 2 to 8°C as supplied. | |
IgG1 |
Flow Cytometry | |
758926 | |
Flow Cytometry 0.25-1 μg/106 cells | |
Q92982 | |
NINJ1 | |
E. coli-derived recombinant human Ninjurin-1, Asp2-Val81, Accession # Q92982 | |
100 μg | |
4814 | |
Human | |
Supernatant |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction