Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human Ninjurin-1 Alexa Fluor 750-conjugated Antibody, R&D Systems™

Mouse Monoclonal Antibody
Supplier: R&D Systems FAB51051S100UG
This item is not returnable.
View return policy
Description
Ninjurin-1 Monoclonal specifically detects Ninjurin-1 in Human samples. It is validated for Flow Cytometry.Specifications
Ninjurin-1 | |
Monoclonal | |
0.2 mg/mL | |
Flow Cytometry 0.25-1 ug/10^6 cells | |
Q92982 | |
NINJ1 | |
E. coli-derived recombinant human Ninjurin-1 Asp2-Val81 Accession # Q92982 | |
RUO | |
4814 | |
Human | |
IgG1 |
Flow Cytometry | |
758926 | |
Alexa Fluor 750 | |
Supplied 0.2 mg/mL in a saline solution containing BSA and Sodium Azide. with 0.09% Sodium Azide | |
Nerve injury-induced protein 1, nerve injury-induced protein-1, NIN1, NINJ1, NINJURIN, ninjurin 1, ninjurin-1, Ninjurin1 | |
Mouse | |
100 μg | |
Primary | |
Detects human Ninjurin-1 in direct ELISAs. In direct ELISAs, no cross-reactivity with recombinant human Ninjurin-2 is observed. | |
Store the unopened product at 2 - 8 degreesC. Do not use past expiration date. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only (RUO)
Spot an opportunity for improvement?Share a Content Correction